missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzenesulfonyl chloride, 97%

Catalog No. AAA1408114
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1408114 25 g
AAA1408103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1408114 Supplier Thermo Scientific Chemicals Supplier No. A1408114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 49584-26-1 | C7H4ClNO2S | 201.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 49584-26-1
Molecular Formula C7H4ClNO2S
Molecular Weight (g/mol) 201.62
MDL Number MFCD00052337
InChI Key DBMFYTQPPBBKHI-UHFFFAOYSA-N
Synonym 4-cyanobenzene-1-sulfonyl chloride, 4-cyanobenzenesulfonylchloride, p-cyanobenzenesulphonyl chloride, 4-cyano benzenesulfonyl chloride, benzenesulfonyl chloride, 4-cyano, 4-cyanophenylsulfonyl chloride, p-cyanobenzenesulfonyl chloride, 4-cyanobenzenesulphonyl chloride, 4-cyano-benzenesulfonyl chloride, 4-chlorosulfonyl benzenecarbonitrile
PubChem CID 39497
SMILES ClS(=O)(=O)C1=CC=C(C=C1)C#N

Specifications

Melting Point 107°C to 110°C
Quantity 25 g
UN Number UN3261
Beilstein 777939
Sensitivity Moisture sensitive
IUPAC Name 4-cyanobenzenesulfonyl chloride
Formula Weight 201.63
Percent Purity 97%
Chemical Name or Material 4-Cyanobenzenesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only