missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzeneboronic acid pinacol ester, 97%

Catalog No. p-7030369
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2820803 1 g
AAH2820806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2820803 Supplier Thermo Scientific Chemicals Supplier No. H2820803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 171364-82-2 | C13H16BNO2 | 229.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 171364-82-2
Molecular Formula C13H16BNO2
Molecular Weight (g/mol) 229.09
MDL Number MFCD03093897
InChI Key HOPDTPGXBZCBNP-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-cyanophenylboronic acid pinacol ester, 4-cyanobenzeneboronic acid pinacol ester, 4-cyanophenylboronic acid, pinacol ester, benzonitrile, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-cyanobenzeneboronic acid, pinacol ester, 2-4-cyanophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 4-cyanophenylboronicacidpinacolester
PubChem CID 2734625
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C#N

Specifications

Quantity 1 g
Formula Weight 229.08
Percent Purity 97%
Chemical Name or Material 4-Cyanobenzeneboronic acid pinacol ester
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only