missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyanobenzamide 97.0+%, TCI America™
$128.98 - $256.05
Chemical Identifiers
| CAS | 3034-34-2 |
|---|---|
| Molecular Formula | C8H6N2O |
| Molecular Weight (g/mol) | 146.15 |
| MDL Number | MFCD00017133 |
| InChI Key | FUKWTMJZHKZKFA-UHFFFAOYSA-N |
| Synonym | p-cyanobenzamide, benzamide, 4-cyano, unii-3p9ug7v78s, benzamide, p-cyano, benzamide, p-cyano-8ci, p-cyano benzamide, 4-cyano-benzamide, cyanobenzamide, acmc-1agnt, p-cyanobenzoic acid amide |
| PubChem CID | 76427 |
| IUPAC Name | 4-cyanobenzamide |
| SMILES | NC(=O)C1=CC=C(C=C1)C#N |
Chemical Identifiers
| 3034-34-2 | |
| 146.15 | |
| FUKWTMJZHKZKFA-UHFFFAOYSA-N | |
| 76427 | |
| NC(=O)C1=CC=C(C=C1)C#N |
| C8H6N2O | |
| MFCD00017133 | |
| p-cyanobenzamide, benzamide, 4-cyano, unii-3p9ug7v78s, benzamide, p-cyano, benzamide, p-cyano-8ci, p-cyano benzamide, 4-cyano-benzamide, cyanobenzamide, acmc-1agnt, p-cyanobenzoic acid amide | |
| 4-cyanobenzamide |
Specifications
| 3034-34-2 | |
| White-Yellow | |
| MFCD00017133 | |
| p-cyanobenzamide, benzamide, 4-cyano, unii-3p9ug7v78s, benzamide, p-cyano, benzamide, p-cyano-8ci, p-cyano benzamide, 4-cyano-benzamide, cyanobenzamide, acmc-1agnt, p-cyanobenzoic acid amide | |
| NC(=O)C1=CC=C(C=C1)C#N | |
| 146.15 | |
| 146.15 | |
| Crystalline Powder |
| 226°C | |
| C8H6N2O | |
| 1 g | |
| FUKWTMJZHKZKFA-UHFFFAOYSA-N | |
| 4-cyanobenzamide | |
| 76427 | |
| ≥97.0% (N) | |
| 4-Cyanobenzamide |
Safety and Handling
RTECSNumber : CV2484500
TSCA : No