missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzamide 97.0+%, TCI America™

Catalog No. C15945G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15945G 5 g
C15941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15945G Supplier TCI America Supplier No. C15945G

Chemical Identifiers

CAS 3034-34-2
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.15
MDL Number MFCD00017133
InChI Key FUKWTMJZHKZKFA-UHFFFAOYSA-N
Synonym p-cyanobenzamide, benzamide, 4-cyano, unii-3p9ug7v78s, benzamide, p-cyano, benzamide, p-cyano-8ci, p-cyano benzamide, 4-cyano-benzamide, cyanobenzamide, acmc-1agnt, p-cyanobenzoic acid amide
PubChem CID 76427
IUPAC Name 4-cyanobenzamide
SMILES NC(=O)C1=CC=C(C=C1)C#N

Specifications

Melting Point 226°C
Color White-Yellow
Quantity 5 g
Formula Weight 146.15
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanobenzamide
RTECSNumber CV2484500
TSCA No