missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyanobenzamide 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 226°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 146.15 |
| Percent Purity | ≥97.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Cyanobenzamide |
Safety and Handling
| RTECSNumber | CV2484500 |
| TSCA | No |