missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzamide, 97%

Catalog No. AAB2560003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2560003 1 g
AAB2560006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2560003 Supplier Thermo Scientific Chemicals Supplier No. B2560003
Only null left

CAS: 3034-34-2 | C8H6N2O | 146.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3034-34-2
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.15
MDL Number MFCD00017133
InChI Key FUKWTMJZHKZKFA-UHFFFAOYSA-N
Synonym p-cyanobenzamide, benzamide, 4-cyano, unii-3p9ug7v78s, benzamide, p-cyano, benzamide, p-cyano-8ci, p-cyano benzamide, 4-cyano-benzamide, cyanobenzamide, acmc-1agnt, p-cyanobenzoic acid amide
PubChem CID 76427
IUPAC Name 4-cyanobenzamide
SMILES NC(=O)C1=CC=C(C=C1)C#N

Specifications

Melting Point 225°C to 228°C
Quantity 1 g
Formula Weight 146.15
Percent Purity 97%
Chemical Name or Material 4-Cyanobenzamide
Hazard Category H302+H312
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
EINECSNumber 221-223-1
RTECSNumber CV2484500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only