missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-L-phenylalanine, 95%

Catalog No. AAH63572MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63572MD 250 mg
AAH6357203 1 g
AAH6357206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63572MD Supplier Thermo Scientific Chemicals Supplier No. H63572MD
Only null left

CAS: 167479-78-9 | C10H10N2O2 | 190.202 g/mol

4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 167479-78-9
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.202
MDL Number MFCD00270363
InChI Key KWIPUXXIFQQMKN-VIFPVBQESA-N
Synonym l-4-cyanophenylalanine, 4-cyano-l-phenylalanine, s-2-amino-3-4-cyanophenyl propanoic acid, h-phe 4-cn-oh, 2s-2-amino-3-4-cyanophenyl propanoic acid, 4-cyanophenylalanine, s-2-amino-3-4-cyanophenyl propionic acid, d-4-cn-phe-oh
PubChem CID 1501865
IUPAC Name (2S)-2-amino-3-(4-cyanophenyl)propanoic acid
SMILES C1=CC(=CC=C1CC(C(=O)O)N)C#N

Specifications

UN Number UN3439
Solubility Information Slightly soluble in water.
Quantity 250 mg
Formula Weight 190.2
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Cyano-L-phenylalanine
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only