missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4-phenylpiperidine hydrochloride, 97%

Catalog No. AAH2841403
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2841403 1 g
AAH2841409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2841403 Supplier Thermo Scientific Chemicals Supplier No. H2841403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 51304-58-6 | C12H15ClN2 | 222.716 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51304-58-6
Molecular Formula C12H15ClN2
Molecular Weight (g/mol) 222.716
MDL Number MFCD00012775
InChI Key CQPHZBOPSZGTJM-UHFFFAOYSA-N
Synonym 4-cyano-4-phenylpiperidine hydrochloride, 4-phenylpiperidine-4-carbonitrile hydrochloride, 4-cyano-4-phenylpiperidinehydrochloride, 4-phenylpiperidine-4-carbonitrile, chloride, 4-phenylpiperidine-4-carbonitrile hcl, pubchem19077, acmc-20a3zw, 4-cyano-4-phenylpiperidine hcl, timtec-bb sbb003334, 4-phenylisonipecotonitrile hydrochloride
PubChem CID 3084973
IUPAC Name 4-phenylpiperidine-4-carbonitrile;hydrochloride
SMILES C1CNCCC1(C#N)C2=CC=CC=C2.Cl

Specifications

Melting Point 210°C to 214°C
Quantity 1 g
UN Number UN3439
Formula Weight 222.71
Percent Purity 97%
Chemical Name or Material 4-Cyano-4-phenylpiperidine hydrochloride
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 257-124-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only