missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-2-fluoropyridine, 97%

CAS: 3939-14-8 | C6H3FN2 | 122.102 g/mol

Supplier:  Thermo Scientific Chemicals H2593814

 View more versions of this product

Catalog No. AAH2593814

Only null left
Explore available promotions

Description

Description

4-Cyano-2-fluoropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-2-fluoropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with hydrogen fluoride, nitrogen oxides, strong acids, strong bases, strong oxidizing agents and strong reducing agents.
Specifications

Specifications

4-Cyano-2-fluoropyridine
31°C to 33°C
MFCD09027299
2-fluoroisonicotinonitrile, 4-cyano-2-fluoropyridine, 2-fluoro-4-cyanopyridine, 4-pyridinecarbonitrile, 2-fluoro, 2-fluoro-4-pyridinecarbonitrile, 2-fluoro-isonicotinonitrile, 4-pyridinecarbonitrile,2-fluoro, acmc-1bnd8, 4-cyano-2-fluoro pyridine, ksc581g3h
DYQMGPSBUBTNJZ-UHFFFAOYSA-N
2-fluoropyridine-4-carbonitrile
12002587
97%
3939-14-8
C6H3FN2
25 g
Insoluble in water.
C1=CN=C(C=C1C#N)F
122.102
122.1

Chemical Identifiers

3939-14-8
122.102
DYQMGPSBUBTNJZ-UHFFFAOYSA-N
2-fluoropyridine-4-carbonitrile
C6H3FN2
MFCD09027299
12002587
C1=CN=C(C=C1C#N)F
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c

H302-H315-H319

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only