Learn More
4-Cyano-2-fluorobenzeneboronic acid, 98%, Thermo Scientific Chemicals
$377.78 - $377.78
Chemical Identifiers
| CAS | 1150114-77-4 |
|---|---|
| Molecular Formula | C7H5BFNO2 |
| Molecular Weight (g/mol) | 164.93 |
| MDL Number | MFCD07644472 |
| InChI Key | HFAUMAIVCYTVQR-UHFFFAOYSA-N |
| Synonym | 4-cyano-2-fluorophenyl boronic acid, boronic acid, b-4-cyano-2-fluorophenyl, acmc-2099nb, 4-borono-3-fluorobenzonitrile, 4-cyano-2-fluoro-phenylboronic acid, 4-cyano-2-fluorobenzeneboronic acid |
| PubChem CID | 44755209 |
| IUPAC Name | (4-cyano-2-fluorophenyl)boronic acid |
| SMILES | OB(O)C1=C(F)C=C(C=C1)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6113403
|
Thermo Scientific Chemicals
H6113403 |
1 g |
Each for $377.78
|
|
|||||
Description
4-Cyano-2-fluorobenzeneboronic acid is used an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Cyano-2-fluorobenzeneboronic acid is used an active pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
| 1150114-77-4 | |
| 164.93 | |
| HFAUMAIVCYTVQR-UHFFFAOYSA-N | |
| 44755209 | |
| OB(O)C1=C(F)C=C(C=C1)C#N |
| C7H5BFNO2 | |
| MFCD07644472 | |
| 4-cyano-2-fluorophenyl boronic acid, boronic acid, b-4-cyano-2-fluorophenyl, acmc-2099nb, 4-borono-3-fluorobenzonitrile, 4-cyano-2-fluoro-phenylboronic acid, 4-cyano-2-fluorobenzeneboronic acid | |
| (4-cyano-2-fluorophenyl)boronic acid |
Specifications
| 1150114-77-4 | |
| MFCD07644472 | |
| 4-cyano-2-fluorophenyl boronic acid, boronic acid, b-4-cyano-2-fluorophenyl, acmc-2099nb, 4-borono-3-fluorobenzonitrile, 4-cyano-2-fluoro-phenylboronic acid, 4-cyano-2-fluorobenzeneboronic acid | |
| OB(O)C1=C(F)C=C(C=C1)C#N | |
| 164.93 | |
| 164.93 | |
| Solid |
| C7H5BFNO2 | |
| 1 g | |
| HFAUMAIVCYTVQR-UHFFFAOYSA-N | |
| (4-cyano-2-fluorophenyl)boronic acid | |
| 44755209 | |
| 98% | |
| 4-Cyano-2-fluorobenzeneboronic acid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only