missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-1-cyclohexene 98.0+%, TCI America™

Catalog No. C08375ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C08375ML 5 mL
C083725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C08375ML Supplier TCI America Supplier No. C08375ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100-45-8
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00013778
InChI Key GYBNBQFUPDFFQX-UHFFFAOYSA-N
Synonym 3-cyclohexene-1-carbonitrile, 4-cyanocyclohexene, 3-cyclohexenecarbonitrile, 4-cyano-1-cyclohexene, cyclohex-3-enecarbonitrile, 3-cyclohexenyl cyanide, 1-cyano-3-cyclohexene, 3-cyklohexenylkyanid, cyclohex-3-enenitrile, 3-cyklohexenylkyanid czech
PubChem CID 66013
IUPAC Name cyclohex-3-ene-1-carbonitrile
SMILES C1CC(CC=C1)C#N

Specifications

Color Yellow
Boiling Point 60°C
Quantity 5 mL
UN Number 3276
Formula Weight 107.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Cyano-1-cyclohexene
RTECSNumber GW2625000
TSCA No