missing translation for 'onlineSavingsMsg'
Learn More

4-Chromanone 98.0+%, TCI America™

Catalog No. C11755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11755G 5 g
C117525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C11755G Supplier TCI America Supplier No. C11755G

Chemical Identifiers

CAS 491-37-2
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00006840
InChI Key MSTDXOZUKAQDRL-UHFFFAOYSA-N
Synonym 4-chromanone, chroman-4-one, chromanone, 4h-1-benzopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-1-benzopyran-4-one, isochromanone, 3,4-dihydro-2h-1-benzopyran-4-one, 2,3-dihydro-4h-1-benzopyran-4-one, dihydrochromone
PubChem CID 68110
IUPAC Name 2,3-dihydrochromen-4-one
SMILES C1COC2=CC=CC=C2C1=O

Specifications

Color White-Yellow
Boiling Point 128°C
Quantity 5 g
Formula Weight 148.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chromanone
RTECSNumber DJ2981225
TSCA Yes
Recommended Storage Refrigerator