missing translation for 'onlineSavingsMsg'
Learn More

4-Chromanol 98.0+%, TCI America™

Catalog No. c16271g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16271G Supplier TCI America Supplier No. C16271G
Only null left

Chemical Identifiers

CAS 1481-93-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00006849
InChI Key MGSHXMOLUWTMGP-UHFFFAOYSA-N
Synonym 4-chromanol, chroman-4-ol, 3,4-dihydro-2h-1-benzopyran-4-ol, 4-hydroxychroman, 2h-1-benzopyran-4-ol, 3,4-dihydro, acmc-20mukj, 3'-carboxy-alpha-chromanol, 2h-1-benzopyran-4-ol,3,4-dihydro
PubChem CID 92890
IUPAC Name 3,4-dihydro-2H-chromen-4-ol
SMILES C1COC2=CC=CC=C2C1O

Specifications

Melting Point 43°C
Boiling Point 80°C
Quantity 1 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chromanol