missing translation for 'onlineSavingsMsg'
Learn More

4-Chromanol, 97%

Catalog No. AAA1494003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1494003 1 g
AAA1494006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1494003 Supplier Thermo Scientific Chemicals Supplier No. A1494003
Only null left

CAS: 1481-93-2 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1481-93-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00006849
InChI Key MGSHXMOLUWTMGP-UHFFFAOYSA-N
PubChem CID 92890
IUPAC Name 3,4-dihydro-2H-chromen-4-ol
SMILES C1COC2=CC=CC=C2C1O

Specifications

Melting Point 38°C to 42°C
Boiling Point 79°C to 81°C (5 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 130800
Synonym 4-chromanol, chroman-4-ol, 3,4-dihydro-2h-1-benzopyran-4-ol, 4-hydroxychroman, 2h-1-benzopyran-4-ol, 3,4-dihydro, acmc-20mukj, 3'-carboxy-alpha-chromanol, 2h-1-benzopyran-4-ol,3,4-dihydro
Formula Weight 150.18
Percent Purity 97%
Chemical Name or Material 4-Chromanol
EINECSNumber 216-041-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only