missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorotoluene 98.0+%, TCI America™

Catalog No. p-7124345
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C029725G 25 g
C0297500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C029725G Supplier TCI America Supplier No. C029725G
Only null left

Chemical Identifiers

CAS 106-43-4
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.58
MDL Number MFCD00000631
InChI Key NPDACUSDTOMAMK-UHFFFAOYSA-N
Synonym 4-chlorotoluene, p-chlorotoluene, benzene, 1-chloro-4-methyl, p-tolyl chloride, toluene, p-chloro, 1-methyl-4-chlorobenzene, 4-chloro-1-methylbenzene, monochlorotoluene, para-chlorotoluene, unii-q8r236h42n
PubChem CID 7810
ChEBI CHEBI:34401
IUPAC Name 1-chloro-4-methylbenzene
SMILES CC1=CC=C(Cl)C=C1

Specifications

Color Colorless
Quantity 25 g
Physical Form Clear Liquid at 20°C
UN Number 2238
Boiling Point 162°C
Formula Weight 126.58
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Chlorotoluene
EINECSNumber (3)-0039
RTECSNumber XS9010000
TSCA Yes