missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorothiobenzamide 97.0+%, TCI America™
Supplier: TCI America C205325G
Specifications
| 4-Chlorothiobenzamide | |
| 2521-24-6 | |
| Crystal-Powder at 20°C | |
| MFCD00040956 | |
| 4-chlorothiobenzamide, 4-chlorobenzothioamide, p-chlorothiobenzamide, p-chlorobenzothiamide, 4-chlorobenzene-1-carbothioamide, 4-chloro-thiobenzamide, benzenecarbothioamide, 4-chloro, benzamide, p-chlorothio, benzamide, p-chlorothio-8ci, amino 4-chlorophenyl methane-1-thione | |
| NC(=S)C1=CC=C(Cl)C=C1 | |
| 171.64 | |
| 171.64 |
| 129°C | |
| 25 g | |
| C7H6ClNS | |
| 2811 | |
| OKPUICCJRDBRJT-UHFFFAOYSA-N | |
| 4-chlorobenzene-1-carbothioamide | |
| 2734826 | |
| ≥97.0% (HPLC,N) |
Chemical Identifiers
| 2521-24-6 | |
| 171.64 | |
| OKPUICCJRDBRJT-UHFFFAOYSA-N | |
| 2734826 | |
| NC(=S)C1=CC=C(Cl)C=C1 |
| C7H6ClNS | |
| MFCD00040956 | |
| 4-chlorothiobenzamide, 4-chlorobenzothioamide, p-chlorothiobenzamide, p-chlorobenzothiamide, 4-chlorobenzene-1-carbothioamide, 4-chloro-thiobenzamide, benzenecarbothioamide, 4-chloro, benzamide, p-chlorothio, benzamide, p-chlorothio-8ci, amino 4-chlorophenyl methane-1-thione | |
| 4-chlorobenzene-1-carbothioamide |
Safety and Handling
TSCA : No