missing translation for 'onlineSavingsMsg'
Learn More

4-chlorotetrahydropyran, 96%

Catalog No. AC434270050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434270050 5 g Glass bottle
AC434270010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434270050 Supplier Thermo Scientific Chemicals Supplier No. 434270050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1768-64-5 | C5H9ClO | 120.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1768-64-5
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
InChI Key DHRSKOBIDIDMJZ-UHFFFAOYSA-N
Synonym 4-chlorotetrahydropyran, 4-chlorotetrahydro-2h-pyran, 2h-pyran, 4-chlorotetrahydro, 4-chloro-tetrahydro-pyran, 4-chloro-tetrahydro-2h-pyran, 4-chloranyloxane, 4-chlorotetrapyran, 4-chloro-tetrahydropyran, acmc-1bvar, bromomethylpentafluorobenzene
PubChem CID 137202
IUPAC Name 4-chlorooxane
SMILES C1COCCC1Cl

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1200g/mL
Boiling Point 150°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.12
Formula Weight 120.58
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 4-chlorotetrahydropyran
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapour.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only