missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorotetrahydro-2H-pyran 98.0+%, TCI America™
Supplier: TCI America C227325G
Specifications
| 4-Chlorotetrahydro-2H-pyran | |
| Yellow | |
| C5H9ClO | |
| 25 g | |
| 4-chlorotetrahydropyran, 4-chlorotetrahydro-2h-pyran, 2h-pyran, 4-chlorotetrahydro, 4-chloro-tetrahydro-pyran, 4-chloro-tetrahydro-2h-pyran, 4-chloranyloxane, 4-chlorotetrapyran, 4-chloro-tetrahydropyran, acmc-1bvar, bromomethylpentafluorobenzene | |
| C1COCCC1Cl | |
| 120.576 | |
| 120.58 | |
| Liquid |
| 1768-64-5 | |
| 150°C | |
| MFCD00074911 | |
| 1993 | |
| DHRSKOBIDIDMJZ-UHFFFAOYSA-N | |
| 4-chlorooxane | |
| 137202 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1768-64-5 | |
| 120.576 | |
| DHRSKOBIDIDMJZ-UHFFFAOYSA-N | |
| 137202 | |
| C1COCCC1Cl |
| C5H9ClO | |
| MFCD00074911 | |
| 4-chlorotetrahydropyran, 4-chlorotetrahydro-2h-pyran, 2h-pyran, 4-chlorotetrahydro, 4-chloro-tetrahydro-pyran, 4-chloro-tetrahydro-2h-pyran, 4-chloranyloxane, 4-chlorotetrapyran, 4-chloro-tetrahydropyran, acmc-1bvar, bromomethylpentafluorobenzene | |
| 4-chlorooxane |
Safety and Handling
TSCA : No