missing translation for 'onlineSavingsMsg'
Learn More

4-(Chlorosulfonyl)benzoic acid, 96%

Catalog No. AAA1733714
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1733714 25 g
AAA1733703 1 g
AAA1733706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1733714 Supplier Thermo Scientific Chemicals Supplier No. A1733714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10130-89-9 | C7H5ClO4S | 220.623 g/mol

It is a chemical reagent used in catalyzing living radical polymerizations, in the preparation of polymer bound transfer hydrogenation catalyst1 and 4-(carboxymethyl-sulfamoyl)-benzoyladenosine. It is also used as a reagent in the sulfonation of γ-cyclodextrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a chemical reagent used in catalyzing living radical polymerizations, in the preparation of polymer bound transfer hydrogenation catalyst1 and 4-(carboxymethyl-sulfamoyl)-benzoyladenosine. It is also used as a reagent in the sulfonation of γ-cyclodextrin.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 10130-89-9
Molecular Formula C7H5ClO4S
Molecular Weight (g/mol) 220.623
MDL Number MFCD00007448
InChI Key PTCSSXYPZOFISK-UHFFFAOYSA-N
Synonym 4-chlorosulfonyl benzoic acid, benzoic acid, 4-chlorosulfonyl, p-chlorosulfonyl benzoic acid, 4-chlorosulphonyl benzoic acid, p-carboxybenzenesulfonyl chloride, benzoic acid, p-chlorosulfonyl, 4-chlorosulfonyl-benzoic acid, 4-carboxybenzenesulfonyl chloride, 4-carboxy-benzenesulfonylchloride, pubchem14811
PubChem CID 82385
IUPAC Name 4-chlorosulfonylbenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)S(=O)(=O)Cl

Specifications

Melting Point 228°C to 232°C
Odor Acrid
Quantity 25 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 220.63
Percent Purity 96%
Chemical Name or Material 4-(Chlorosulfonyl)benzoic acid
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 233-367-2
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only