missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorosalicylaldehyde 95.0+%, TCI America™
$145.40 - $577.86
Chemical Identifiers
| CAS | 2420-26-0 |
|---|---|
| Molecular Formula | C7H5ClO2 |
| Molecular Weight (g/mol) | 156.57 |
| MDL Number | MFCD06252499 |
| InChI Key | QNZWAJZEJAOVPN-UHFFFAOYSA-N |
| Synonym | 4-Chloro-2-hydroxybenzaldehyde |
| PubChem CID | 520101 |
| IUPAC Name | 4-chloro-2-hydroxybenzaldehyde |
| SMILES | OC1=C(C=O)C=CC(Cl)=C1 |
Chemical Identifiers
| 2420-26-0 | |
| 156.57 | |
| QNZWAJZEJAOVPN-UHFFFAOYSA-N | |
| 520101 | |
| OC1=C(C=O)C=CC(Cl)=C1 |
| C7H5ClO2 | |
| MFCD06252499 | |
| 4-Chloro-2-hydroxybenzaldehyde | |
| 4-chloro-2-hydroxybenzaldehyde |
Specifications
| 2420-26-0 | |
| C7H5ClO2 | |
| 1 g | |
| QNZWAJZEJAOVPN-UHFFFAOYSA-N | |
| 4-chloro-2-hydroxybenzaldehyde | |
| 520101 | |
| ≥95.0% (GC,T) | |
| 4-Chlorosalicylaldehyde |
| 47°C | |
| MFCD06252499 | |
| 4-Chloro-2-hydroxybenzaldehyde | |
| OC1=C(C=O)C=CC(Cl)=C1 | |
| 156.57 | |
| 156.57 | |
| Crystalline Powder |
Safety and Handling
TSCA : No