missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroquinoline 97.0+%, TCI America™

Catalog No. C22841G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22841G 1 g
C22845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22841G Supplier TCI America Supplier No. C22841G

Chemical Identifiers

CAS 611-35-8
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.60
MDL Number MFCD00006773
InChI Key KNDOFJFSHZCKGT-UHFFFAOYSA-N
Synonym quinoline, 4-chloro, 4-chloro-quinoline, ccris 3979, 4-chioroquinoline, acmc-209mpi, 4-chloroquinoline, ksc354e5n, 4-chloroquinoline 1g, kndofjfshzckgt-uhfffaoysa
PubChem CID 69140
IUPAC Name 4-chloroquinoline
SMILES ClC1=C2C=CC=CC2=NC=C1

Specifications

Color White-Yellow
Boiling Point 261°C
Quantity 1 g
Formula Weight 163.60
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Chloroquinoline
TSCA No
Recommended Storage Refrigerator