missing translation for 'onlineSavingsMsg'
Learn More

4-chloroquinoline, 99%

Catalog No. AC110050010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC110050010 1 g
AC110050050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC110050010 Supplier Thermo Scientific Chemicals Supplier No. 110050010

CAS: 611-35-8 | C9H6ClN | 163.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-35-8
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.60
MDL Number MFCD00006773
InChI Key KNDOFJFSHZCKGT-UHFFFAOYSA-N
Synonym quinoline, 4-chloro, 4-chloro-quinoline, ccris 3979, 4-chioroquinoline, acmc-209mpi, 4-chloroquinoline, ksc354e5n, 4-chloroquinoline 1g, kndofjfshzckgt-uhfffaoysa
PubChem CID 69140
IUPAC Name 4-chloroquinoline
SMILES ClC1=C2C=CC=CC2=NC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 29.0°C to 32.0°C
Density 1.2500g/mL
Boiling Point 260.0°C to 261.0°C
Flash Point 110°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 20,36
Specific Gravity 1.25
Formula Weight 163.61
Percent Purity 99%
Chemical Name or Material 4-chloroquinoline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-267-7

RUO – Research Use Only