missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroquinoline-6-boronic acid pinacol ester, 96%

Catalog No. p-7035864
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54085MD 250 mg
AAH5408503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH54085MD Supplier Thermo Scientific Chemicals Supplier No. H54085MD
Only null left

CAS: 1201844-73-6 | C15H17BClNO2 | 289.566 g/mol

4-Chloroquinoline-6-boronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloroquinoline-6-boronic acid pinacol ester is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1201844-73-6
Molecular Formula C15H17BClNO2
Molecular Weight (g/mol) 289.566
MDL Number MFCD16987803
InChI Key ANPKKHFSNJKOAM-UHFFFAOYSA-N
Synonym 4-chloro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 4-chloroquinoline-6-boronic acid pinacol ester, quinoline, 4-chloro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 53363436
IUPAC Name 4-chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=CN=C3C=C2)Cl

Specifications

Quantity 250 mg
Formula Weight 289.56
Percent Purity 96%
Chemical Name or Material 4-Chloroquinoline-6-boronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only