missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloropyridine N-Oxide 97.0+%, TCI America™
$326.21 - $1028.95
Chemical Identifiers
| CAS | 1121-76-2 |
|---|---|
| Molecular Formula | C5H4ClNO |
| Molecular Weight (g/mol) | 129.543 |
| MDL Number | MFCD00047425 |
| InChI Key | DPJVRASYWYOFSJ-UHFFFAOYSA-N |
| Synonym | 4-chloropyridine n-oxide, 4-chloropyridine 1-oxide, 4-chloropyridine oxide, pyridine, 4-chloro-, 1-oxide, 4-chloropyridine-n-oxide, 4-chloro-pyridine 1-oxide, 4-chloropuridine n-oxide, unii-f3196ueb9q, 4-chloropyridin-1-ol, pubchem19817 |
| PubChem CID | 70724 |
| IUPAC Name | 4-chloro-1-oxidopyridin-1-ium |
| SMILES | C1=C[N+](=CC=C1Cl)[O-] |
Chemical Identifiers
| 1121-76-2 | |
| 129.543 | |
| DPJVRASYWYOFSJ-UHFFFAOYSA-N | |
| 70724 | |
| C1=C[N+](=CC=C1Cl)[O-] |
| C5H4ClNO | |
| MFCD00047425 | |
| 4-chloropyridine n-oxide, 4-chloropyridine 1-oxide, 4-chloropyridine oxide, pyridine, 4-chloro-, 1-oxide, 4-chloropyridine-n-oxide, 4-chloro-pyridine 1-oxide, 4-chloropuridine n-oxide, unii-f3196ueb9q, 4-chloropyridin-1-ol, pubchem19817 | |
| 4-chloro-1-oxidopyridin-1-ium |
Specifications
| 1121-76-2 | |
| MFCD00047425 | |
| 4-chloropyridine n-oxide, 4-chloropyridine 1-oxide, 4-chloropyridine oxide, pyridine, 4-chloro-, 1-oxide, 4-chloropyridine-n-oxide, 4-chloro-pyridine 1-oxide, 4-chloropuridine n-oxide, unii-f3196ueb9q, 4-chloropyridin-1-ol, pubchem19817 | |
| C1=C[N+](=CC=C1Cl)[O-] | |
| 129.543 | |
| 129.54 | |
| Crystalline Powder |
| C5H4ClNO | |
| 5 g | |
| DPJVRASYWYOFSJ-UHFFFAOYSA-N | |
| 4-chloro-1-oxidopyridin-1-ium | |
| 70724 | |
| ≥97.0% (GC) | |
| 4-Chloropyridine N-Oxide |
Safety and Handling
TSCA : No