missing translation for 'onlineSavingsMsg'
Learn More

4-Chloropyridine N-Oxide 97.0+%, TCI America™

Catalog No. C15405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15405G 5 g
C154025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15405G Supplier TCI America Supplier No. C15405G

missing translation for 'validMainframeMsg'

CAS: 1121-76-2 | C5H4ClNO | 129.543 g/mol

Chemical Identifiers

CAS 1121-76-2
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.543
MDL Number MFCD00047425
InChI Key DPJVRASYWYOFSJ-UHFFFAOYSA-N
Synonym 4-chloropyridine n-oxide, 4-chloropyridine 1-oxide, 4-chloropyridine oxide, pyridine, 4-chloro-, 1-oxide, 4-chloropyridine-n-oxide, 4-chloro-pyridine 1-oxide, 4-chloropuridine n-oxide, unii-f3196ueb9q, 4-chloropyridin-1-ol, pubchem19817
PubChem CID 70724
IUPAC Name 4-chloro-1-oxidopyridin-1-ium
SMILES C1=C[N+](=CC=C1Cl)[O-]

Specifications

Quantity 5 g
Formula Weight 129.54
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloropyridine N-Oxide