missing translation for 'onlineSavingsMsg'
Learn More

4-Chloropyridine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7124331
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C028125G 25 g
C0281250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C028125G Supplier TCI America Supplier No. C028125G
Only null left

Chemical Identifiers

CAS 7379-35-3
Molecular Formula C5H5Cl2N
Molecular Weight (g/mol) 150.00
MDL Number MFCD00012829
InChI Key XGAFCCUNHIMIRV-UHFFFAOYSA-N
Synonym 4-chloropyridine hydrochloride, 4-chloropyridine hcl, 4-chloropyridinium chloride, pyridine, 4-chloro-, hydrochloride, 4-chloro-pyridine hydrochloride, ccris 1716, pyridine, 4-chloro-, hydrochloride 1:1, 4-chloropyridine, chloride, 4-chloropyridinehydrochloride, 4-chloropyrodine hydrochloride
PubChem CID 81852
IUPAC Name hydrogen 4-chloropyridine chloride
SMILES [H+].[Cl-].ClC1=CC=NC=C1

Specifications

Quantity 25 g
UN Number 2811
Formula Weight 150.00
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloropyridine Hydrochloride