missing translation for 'onlineSavingsMsg'
Learn More

4-Chloropyridine hydrochloride, 98+%

Catalog No. AAA1562806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1562806 5 g
AAA1562814 25 g
AAA1562822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1562806 Supplier Thermo Scientific Chemicals Supplier No. A1562806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7379-35-3 | C5H5Cl2N | 150.00 g/mol

It is employed as a catalyst in the synthesis of fluorazone derivatives by one-pot pyrrolation/cyclization of anthranilic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a catalyst in the synthesis of fluorazone derivatives by one-pot pyrrolation/cyclization of anthranilic acids.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 7379-35-3
Molecular Formula C5H5Cl2N
Molecular Weight (g/mol) 150.00
MDL Number MFCD00012829
InChI Key XGAFCCUNHIMIRV-UHFFFAOYSA-N
Synonym 4-chloropyridine hydrochloride, 4-chloropyridine hcl, 4-chloropyridinium chloride, pyridine, 4-chloro-, hydrochloride, 4-chloro-pyridine hydrochloride, ccris 1716, pyridine, 4-chloro-, hydrochloride 1:1, 4-chloropyridine, chloride, 4-chloropyridinehydrochloride, 4-chloropyrodine hydrochloride
PubChem CID 81852
IUPAC Name 4-chloropyridine;hydrochloride
SMILES [H+].[Cl-].ClC1=CC=NC=C1

Specifications

Melting Point ∼210°C (sublimation)
pH 2
Flash Point 198°C (388°F)
Odor Weak
Quantity 5 g
Beilstein 3684904
Solubility Information Soluble in water.
Formula Weight 150.01
Percent Purity ≥98%
Chemical Name or Material 4-Chloropyridine hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 230-946-1
RTECSNumber US6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only