missing translation for 'onlineSavingsMsg'
Learn More

4-Chloropyridine-2-carboxylic acid, 94%

Catalog No. p-7031285
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3230103 1 g
AAH3230109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3230103 Supplier Thermo Scientific Chemicals Supplier No. H3230103
Only null left

CAS: 5470-22-4 | C6H4ClNO2 | 157.553 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5470-22-4
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.553
MDL Number MFCD00191400
InChI Key NNMYRMGMVLMQAY-UHFFFAOYSA-N
Synonym 4-chloropicolinic acid, 4-chloro-pyridine-2-carboxylic acid, 4-chloro-2-pyridinecarboxylic acid, 4-chloro-2-picolinic acid, 2-pyridinecarboxylic acid, 4-chloro, 4-chloro picolinic acid, 2-carboxy-4-chloropyridine, chloropicolinicacid, pubchem12731, acmc-1bn7z
PubChem CID 230890
IUPAC Name 4-chloropyridine-2-carboxylic acid
SMILES C1=CN=C(C=C1Cl)C(=O)O

Specifications

Melting Point ∽180°C (decomposition)
Quantity 1 g
Formula Weight 157.56
Percent Purity 94%
Chemical Name or Material 4-Chloropyridine-2-carboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only