missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloropyridine-2-carboxamide 97.0+%, TCI America™
$33.56 - $128.62
Chemical Identifiers
| CAS | 99586-65-9 |
|---|---|
| Molecular Formula | C6H5ClN2O |
| Molecular Weight (g/mol) | 156.569 |
| MDL Number | MFCD01085339 |
| InChI Key | XIHHOUUTBZSYJH-UHFFFAOYSA-N |
| Synonym | 4-chloropicolinamide, 4-chloro-pyridine-2-carboxylic acid amide, 4-chloro-2-pyridinecarboxamide, 4-chlorophcolinamide, 4-chloropyridine-2-carboxylic acid amide, 2-pyridinecarboxamide, 4-chloro, 4-chloro-pyridine-2-carboxylicacidamide, pubchem9223, acmc-209sda, ksc486q9h |
| PubChem CID | 303543 |
| IUPAC Name | 4-chloropyridine-2-carboxamide |
| SMILES | C1=CN=C(C=C1Cl)C(=O)N |
Chemical Identifiers
| 99586-65-9 | |
| 156.569 | |
| XIHHOUUTBZSYJH-UHFFFAOYSA-N | |
| 303543 | |
| C1=CN=C(C=C1Cl)C(=O)N |
| C6H5ClN2O | |
| MFCD01085339 | |
| 4-chloropicolinamide, 4-chloro-pyridine-2-carboxylic acid amide, 4-chloro-2-pyridinecarboxamide, 4-chlorophcolinamide, 4-chloropyridine-2-carboxylic acid amide, 2-pyridinecarboxamide, 4-chloro, 4-chloro-pyridine-2-carboxylicacidamide, pubchem9223, acmc-209sda, ksc486q9h | |
| 4-chloropyridine-2-carboxamide |
Specifications
| 99586-65-9 | |
| White-Yellow | |
| MFCD01085339 | |
| 4-chloropicolinamide, 4-chloro-pyridine-2-carboxylic acid amide, 4-chloro-2-pyridinecarboxamide, 4-chlorophcolinamide, 4-chloropyridine-2-carboxylic acid amide, 2-pyridinecarboxamide, 4-chloro, 4-chloro-pyridine-2-carboxylicacidamide, pubchem9223, acmc-209sda, ksc486q9h | |
| C1=CN=C(C=C1Cl)C(=O)N | |
| 156.569 | |
| 156.57 | |
| Crystalline Powder |
| 162°C | |
| C6H5ClN2O | |
| 1 g | |
| XIHHOUUTBZSYJH-UHFFFAOYSA-N | |
| 4-chloropyridine-2-carboxamide | |
| 303543 | |
| ≥97.0% (HPLC,T) | |
| 4-Chloropyridine-2-carboxamide |
Safety and Handling
TSCA : No