missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloropivaloanilide 98.0+%, TCI America™

Catalog No. C209925G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C209925G 25 g
C2099250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C209925G Supplier TCI America Supplier No. C209925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 65854-91-3
Molecular Formula C11H14ClNO
Molecular Weight (g/mol) 211.689
MDL Number MFCD01571825
InChI Key IZISMXMXCLUHGI-UHFFFAOYSA-N
Synonym 4-Chloro-N-pivaloylaniline, N-(4-Chlorophenyl)pivalamide, N-(4-Chlorophenyl)-2,2-dimethylpropionamide
PubChem CID 182511
IUPAC Name N-(4-chlorophenyl)-2,2-dimethylpropanamide
SMILES CC(C)(C)C(=O)NC1=CC=C(C=C1)Cl

Specifications

Melting Point 152°C
Color White-Yellow
Quantity 25 g
Formula Weight 211.69
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chloropivaloanilide