missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophthalic Anhydride 98.0+%, TCI America™

Catalog No. c13285g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C13285G 5 g
C132825G 25 g
2 options

Catalog No. C13285G

Supplier: TCI America C13285G

Only null left

Chemical Identifiers

CAS 118-45-6
Molecular Formula C8H3ClO3
Molecular Weight (g/mol) 182.56
MDL Number MFCD00152354
InChI Key BTTRMCQEPDPCPA-UHFFFAOYSA-N
PubChem CID 67044
IUPAC Name 5-chloro-1,3-dihydro-2-benzofuran-1,3-dione
SMILES ClC1=CC=C2C(=O)OC(=O)C2=C1

Specifications

Melting Point 96°C
Color White
Boiling Point 290°C
Quantity 5 g
Formula Weight 182.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorophthalic Anhydride
EINECSNumber (3)-0062
TSCA No