missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorophenylboronic acid pinacol ester, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Nom ou substance chimique | 4-Chlorophenylboronic acid pinacol ester |
| Point de fusion | 55.0°C to 56.0°C |
| Quantité | 5 g, 5g |
| Point d’éclair | >110°C |
| Emballage | Glass bottle |
| Fourchette de pourcentages d’essai | 96% min. (GC) |
| Synonyme | 2-4-chlorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-chlorophenylboronic acid pinacol ester, 4-chlorophenylboronic acid, pinacol ester, 1,3,2-dioxaborolane,2-4-chlorophenyl-4,4,5,5-tetramethyl, 1,3,2-dioxaborolane, 2-4-chlorophenyl-4,4,5,5-tetramethyl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl chlorobenzene, 4-chlorophenyl boronic acid pinacol ester, 4-chlorobenzeneboronic acid, pinacol ester, p-chlorophenylboronic acid 2,3-dimethyl-2,3-butanediyl |
| Poids de formule | 238.52 |
| Pourcentage de pureté | 97% |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for |
RUO – Research Use Only