missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$94.33 - $94.33
Specifications
| CAS | 1679-18-1 |
|---|---|
| Physical Form | Crystal-Powder at 20°C |
| Molecular Formula | C6H6BClO2 |
| MDL Number | MFCD00039137 |
| InChI Key | CAYQIZIAYYNFCS-UHFFFAOYSA-N |
Specifications
| 1679-18-1 | |
| C6H6BClO2 | |
| CAYQIZIAYYNFCS-UHFFFAOYSA-N | |
| 74299 |
| Crystal-Powder at 20°C | |
| MFCD00039137 | |
| (4-chlorophenyl)boronic acid | |
| 156.37 |