missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorophenylacetyl Chloride 98.0+%, TCI America™
$136.69 - $136.69
Chemical Identifiers
| CAS | 25026-34-0 |
|---|---|
| Molecular Formula | C8H6Cl2O |
| Molecular Weight (g/mol) | 189.04 |
| MDL Number | MFCD00037111 |
| InChI Key | UMQUIRYNOVNYPA-UHFFFAOYSA-N |
| Synonym | 4-chlorophenylacetyl chloride, p-chlorophenylacetyl chloride, 2-4-chlorophenyl acetyl chloride, 4-chlorobenzeneacetyl chloride, benzeneacetyl chloride, 4-chloro, 4-chlorophenylacetylchloride, acmc-1cch1, 4-chlorobenzeneacetylchloride, 4-chlorobenzenacetyl chloride, p-chlorophenyl-acetyl chloride |
| PubChem CID | 90692 |
| IUPAC Name | 2-(4-chlorophenyl)acetyl chloride |
| SMILES | ClC(=O)CC1=CC=C(Cl)C=C1 |
Chemical Identifiers
| 25026-34-0 | |
| 189.04 | |
| UMQUIRYNOVNYPA-UHFFFAOYSA-N | |
| 90692 | |
| ClC(=O)CC1=CC=C(Cl)C=C1 |
| C8H6Cl2O | |
| MFCD00037111 | |
| 4-chlorophenylacetyl chloride, p-chlorophenylacetyl chloride, 2-4-chlorophenyl acetyl chloride, 4-chlorobenzeneacetyl chloride, benzeneacetyl chloride, 4-chloro, 4-chlorophenylacetylchloride, acmc-1cch1, 4-chlorobenzeneacetylchloride, 4-chlorobenzenacetyl chloride, p-chlorophenyl-acetyl chloride | |
| 2-(4-chlorophenyl)acetyl chloride |
Specifications
| 25026-34-0 | |
| 129°C | |
| Clear Liquid at 20°C | |
| MFCD00037111 | |
| 4-chlorophenylacetyl chloride, p-chlorophenylacetyl chloride, 2-4-chlorophenyl acetyl chloride, 4-chlorobenzeneacetyl chloride, benzeneacetyl chloride, 4-chloro, 4-chlorophenylacetylchloride, acmc-1cch1, 4-chlorobenzeneacetylchloride, 4-chlorobenzenacetyl chloride, p-chlorophenyl-acetyl chloride | |
| ClC(=O)CC1=CC=C(Cl)C=C1 | |
| 189.04 | |
| 189.04 | |
| 4-Chlorophenylacetyl Chloride |
| Yellow | |
| 25 g | |
| C8H6Cl2O | |
| 3265 | |
| UMQUIRYNOVNYPA-UHFFFAOYSA-N | |
| 2-(4-chlorophenyl)acetyl chloride | |
| 90692 | |
| ≥98.0% (GC,T) |
Safety and Handling
TSCA : No