Learn More
4-Chlorophenylacetyl chloride, 98%
CAS: 25026-34-0 | C8H6Cl2O | 189.04 g/mol
Supplier: Thermo Scientific Chemicals A1253018
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Chlorophenylacetyl chloride | |
| 1.292 | |
| 50 g | |
| C8H6Cl2O | |
| UN3265 | |
| 4-chlorophenylacetyl chloride, p-chlorophenylacetyl chloride, 2-4-chlorophenyl acetyl chloride, 4-chlorobenzeneacetyl chloride, benzeneacetyl chloride, 4-chloro, 4-chlorophenylacetylchloride, acmc-1cch1, 4-chlorobenzeneacetylchloride, 4-chlorobenzenacetyl chloride, p-chlorophenyl-acetyl chloride | |
| ClC(=O)CC1=CC=C(Cl)C=C1 | |
| 189.04 | |
| 189.04 |
| 25026-34-0 | |
| 112°C to 114°C (11 mmHg) | |
| 1.551 | |
| MFCD00037111 | |
| 637775 | |
| UMQUIRYNOVNYPA-UHFFFAOYSA-N | |
| 2-(4-chlorophenyl)acetyl chloride | |
| 90692 | |
| 98% |
Chemical Identifiers
| 25026-34-0 | |
| 189.04 | |
| UMQUIRYNOVNYPA-UHFFFAOYSA-N | |
| 90692 | |
| ClC(=O)CC1=CC=C(Cl)C=C1 |
| C8H6Cl2O | |
| MFCD00037111 | |
| 4-chlorophenylacetyl chloride, p-chlorophenylacetyl chloride, 2-4-chlorophenyl acetyl chloride, 4-chlorobenzeneacetyl chloride, benzeneacetyl chloride, 4-chloro, 4-chlorophenylacetylchloride, acmc-1cch1, 4-chlorobenzeneacetylchloride, 4-chlorobenzenacetyl chloride, p-chlorophenyl-acetyl chloride | |
| 2-(4-chlorophenyl)acetyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H302-H314-H317
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 246-571-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only