missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylacetyl chloride, 98%

Catalog No. AAA1253009
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1253009 10 g
AAA1253018 50 g
AAA1253030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1253009 Supplier Thermo Scientific Chemicals Supplier No. A1253009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 25026-34-0 | C8H6Cl2O | 189.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25026-34-0
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00037111
InChI Key UMQUIRYNOVNYPA-UHFFFAOYSA-N
Synonym 4-chlorophenylacetyl chloride, p-chlorophenylacetyl chloride, 2-4-chlorophenyl acetyl chloride, 4-chlorobenzeneacetyl chloride, benzeneacetyl chloride, 4-chloro, 4-chlorophenylacetylchloride, acmc-1cch1, 4-chlorobenzeneacetylchloride, 4-chlorobenzenacetyl chloride, p-chlorophenyl-acetyl chloride
PubChem CID 90692
IUPAC Name 2-(4-chlorophenyl)acetyl chloride
SMILES ClC(=O)CC1=CC=C(Cl)C=C1

Specifications

Density 1.292
Quantity 10 g
Refractive Index 1.551
UN Number UN3265
Beilstein 637775
Boiling Point 112°C to 114°C (11 mmHg)
Formula Weight 189.04
Percent Purity 98%
Chemical Name or Material 4-Chlorophenylacetyl chloride
Hazard Category H302-H314-H317
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 246-571-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only