missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylacetyl chloride, 96%

Catalog No. p-85491
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC123450050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC123450050 Supplier Thermo Scientific Chemicals Supplier No. 123450050

CAS: 25026-34-0 | C8H6Cl2O | 189.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25026-34-0
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00037111
InChI Key UMQUIRYNOVNYPA-UHFFFAOYSA-N
Synonym 4-chlorophenylacetyl chloride, p-chlorophenylacetyl chloride, 2-4-chlorophenyl acetyl chloride, 4-chlorobenzeneacetyl chloride, benzeneacetyl chloride, 4-chloro, 4-chlorophenylacetylchloride, acmc-1cch1, 4-chlorobenzeneacetylchloride, 4-chlorobenzenacetyl chloride, p-chlorophenyl-acetyl chloride
PubChem CID 90692
IUPAC Name 2-(4-chlorophenyl)acetyl chloride
SMILES ClC(=O)CC1=CC=C(Cl)C=C1

Specifications

Color Brown to Yellow
CAS Min % 95.0
CAS Max % 100.0
Density 1.2920g/mL
Quantity 5 g
Flash Point >110°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5495 to 1.5565
Specific Gravity 1.292
Assay Percent Range 95% min. (GC)
Linear Formula ClC6H4CH2COCl
Boiling Point 85.0°C (1.0 mmHg)
Solubility Information Solubility in water: reacts.
Formula Weight 189.04
Percent Purity 96%
Chemical Name or Material 4-Chlorophenylacetyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 246-571-1

RUO – Research Use Only