missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylacetic Acid 97.0+%, TCI America™

Catalog No. p-7124682
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C091425G 25 g
C0914500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C091425G Supplier TCI America Supplier No. C091425G
Only null left

Chemical Identifiers

CAS 1878-66-6
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00004344
InChI Key CDPKJZJVTHSESZ-UHFFFAOYSA-N
Synonym 4-chlorophenylacetic acid, 2-4-chlorophenyl acetic acid, 4-chlorophenyl acetic acid, 4-chlorobenzeneacetic acid, p-chlorophenylacetic acid, benzeneacetic acid, 4-chloro, p-chlorophenyl acetic acid, 2-p-chlorophenyl acetic acid, acetic acid, p-chlorophenyl
PubChem CID 15880
ChEBI CHEBI:30749
IUPAC Name 2-(4-chlorophenyl)acetic acid
SMILES OC(=O)CC1=CC=C(Cl)C=C1

Specifications

Melting Point 105°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 170.59
Percent Purity ≥97.0% (T)
Chemical Name or Material 4-Chlorophenylacetic Acid
EINECSNumber (3)-3081
RTECSNumber AG0590000
TSCA Yes