missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylacetic acid, 98%

Catalog No. p-7022278
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1497418 50 g
AAA1497430 250 g
AAA149740B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1497418 Supplier Thermo Scientific Chemicals Supplier No. A1497418

CAS: 1878-66-6 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1878-66-6
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00004344
InChI Key CDPKJZJVTHSESZ-UHFFFAOYSA-N
Synonym 4-chlorophenylacetic acid, 2-4-chlorophenyl acetic acid, 4-chlorophenyl acetic acid, 4-chlorobenzeneacetic acid, p-chlorophenylacetic acid, benzeneacetic acid, 4-chloro, p-chlorophenyl acetic acid, 2-p-chlorophenyl acetic acid, acetic acid, p-chlorophenyl
PubChem CID 15880
ChEBI CHEBI:30749
IUPAC Name 2-(4-chlorophenyl)acetic acid
SMILES OC(=O)CC1=CC=C(Cl)C=C1

Specifications

Melting Point 103°C to 107°C
Quantity 50 g
Odor Odorless
Beilstein 1072816
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material 4-Chlorophenylacetic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 217-521-6
RTECSNumber AG0590000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only