missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylacetic acid, 98%

CAS: 1878-66-6 | C8H7ClO2 | 170.59 g/mol

$368.88 - $368.88

Chemical Identifiers

CAS 1878-66-6
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00004344
InChI Key CDPKJZJVTHSESZ-UHFFFAOYSA-N
Synonym 4-chlorophenylacetic acid, 2-4-chlorophenyl acetic acid, 4-chlorophenyl acetic acid, 4-chlorobenzeneacetic acid, p-chlorophenylacetic acid, benzeneacetic acid, 4-chloro, p-chlorophenyl acetic acid, 2-p-chlorophenyl acetic acid, acetic acid, p-chlorophenyl
PubChem CID 15880
ChEBI CHEBI:30749
IUPAC Name 2-(4-chlorophenyl)acetic acid
SMILES OC(=O)CC1=CC=C(Cl)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC160725000
SDS View Documents
Thermo Scientific Chemicals
160725000
500 g Plastic bottle
Each for $368.88
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1878-66-6
170.59
CDPKJZJVTHSESZ-UHFFFAOYSA-N
15880
2-(4-chlorophenyl)acetic acid
C8H7ClO2
MFCD00004344
4-chlorophenylacetic acid, 2-4-chlorophenyl acetic acid, 4-chlorophenyl acetic acid, 4-chlorobenzeneacetic acid, p-chlorophenylacetic acid, benzeneacetic acid, 4-chloro, p-chlorophenyl acetic acid, 2-p-chlorophenyl acetic acid, acetic acid, p-chlorophenyl
CHEBI:30749
OC(=O)CC1=CC=C(Cl)C=C1

Specifications

103°C to 107°C
White to Cream
100.0
Authentic
Powder
C8H7ClO2
MFCD00004344
4-chlorophenylacetic acid, 2-4-chlorophenyl acetic acid, 4-chlorophenyl acetic acid, 4-chlorobenzeneacetic acid, p-chlorophenylacetic acid, benzeneacetic acid, 4-chloro, p-chlorophenyl acetic acid, 2-p-chlorophenyl acetic acid, acetic acid, p-chlorophenyl
CDPKJZJVTHSESZ-UHFFFAOYSA-N
2-(4-chlorophenyl)acetic acid
15880
170.6
4-Chlorophenylacetic acid, 98%
1878-66-6
97.5
500 g
Plastic bottle
97.5% min. (HPLC)
ClC6H4CH2CO2H
09,448
Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,diethyl ether,solubility 5% in chcl3: clear solution
OC(=O)CC1=CC=C(Cl)C=C1
170.59
CHEBI:30749
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 217-521-6

RTECSNumber : AG0590000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only