missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl Isothiocyanate 98.0+%, TCI America™

Catalog No. I016725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I016725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I016725G Supplier TCI America Supplier No. I016725G

Chemical Identifiers

CAS 2131-55-7
Molecular Formula C7H4ClNS
Molecular Weight (g/mol) 169.626
MDL Number MFCD00004810
InChI Key MZZVFXMTZTVUFO-UHFFFAOYSA-N
Synonym 4-chlorophenyl isothiocyanate, p-chlorophenyl isothiocyanate, 1-chloro-4-isothiocyanato-benzene, benzene, 1-chloro-4-isothiocyanato, 4-chlorophenylisothiocyanate, isothiocyanic acid, p-chlorophenyl ester, 4-chlor-phenyl-isothiocyanat, isothiocyanic acid 4-chlorophenyl ester, isothiocyanic acid, 4-chlorophenyl ester, benzene,1-chloro-4-isothiocyanato
PubChem CID 16480
IUPAC Name 1-chloro-4-isothiocyanatobenzene
SMILES C1=CC(=CC=C1N=C=S)Cl

Specifications

Melting Point 46°C
Color White-Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
UN Number 2811
Boiling Point 127°C
Formula Weight 169.63
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Chlorophenyl Isothiocyanate
RTECSNumber NX8473000
TSCA No