missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl isocyanate, 98%

Catalog No. p-7022272
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1496606 5 g
AAA1496614 25 g
AAA1496622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1496606 Supplier Thermo Scientific Chemicals Supplier No. A1496606
Only null left

CAS: 104-12-1 | C7H4ClNO | 153.565 g/mol

4-Chlorophenyl isocyanate is used in the preparation of isothiocyanates. Aryl isocyanates undergo [2+2] cycloaddition reactions with dihydrofuran to give bicyclic β-lactams in high yield.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorophenyl isocyanate is used in the preparation of isothiocyanates. Aryl isocyanates undergo [2+2] cycloaddition reactions with dihydrofuran to give bicyclic ß-lactams in high yield.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 104-12-1
Molecular Formula C7H4ClNO
Molecular Weight (g/mol) 153.565
MDL Number MFCD00002024
InChI Key ADAKRBAJFHTIEW-UHFFFAOYSA-N
Synonym 4-chlorophenyl isocyanate, p-chlorophenyl isocyanate, benzene, 1-chloro-4-isocyanato, pcpi, 4-chloroisocyanatobenzene, para-chlorophenyl isocyanate, p-chlorfenylisokyanat, 4-chlorophenylisocyanate, isocyanic acid, p-chlorophenyl ester, 1-chloro-4-isocyanato-benzene
PubChem CID 7693
IUPAC Name 1-chloro-4-isocyanatobenzene
SMILES C1=CC(=CC=C1N=C=O)Cl

Specifications

Melting Point 29°C to 31°C
Density 1.2
Quantity 5 g
Odor Pungent
Refractive Index 1.5618
UN Number UN2811
Beilstein 386235
Solubility Information Hydrolyzes in water.
Boiling Point 203°C to 204°C
Formula Weight 153.57
Percent Purity 98%
Chemical Name or Material 4-Chlorophenyl isocyanate
Show More Show Less
Hazard Category H301-H315-H318-H330-H334-H335
Hazard Statement GHS H Statement
H330-H334-H318-H315-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 203-176-9
RTECSNumber NQ8575000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only