missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorophenyl Benzoate 99.0+%, TCI America™
Supplier: TCI America B006325G
Specifications
| 4-Chlorophenyl Benzoate | |
| 89°C | |
| C13H9ClO2 | |
| 25 g | |
| JKSIXXOEIXUYFW-UHFFFAOYSA-N | |
| (4-chlorophenyl) benzoate | |
| 347761 | |
| ≥99.0% (GC) |
| 8-5-2005 | |
| White | |
| MFCD00040854 | |
| benzoic acid, 4-chlorophenyl ester, p-chlorophenol benzoate, p-chlorophenyl benzoate, benzoic acid 4-chlorophenyl ester, benzoic acid, p-chlorophenyl ester, 4-chlorophenylbenzoate, acmc-1cina, cbmicro_005244, 4-chlorophenyl benzoate | |
| C1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)Cl | |
| 232.66 | |
| 232.66 | |
| Crystalline Powder |
Chemical Identifiers
| 8-5-2005 | |
| 232.66 | |
| JKSIXXOEIXUYFW-UHFFFAOYSA-N | |
| 347761 | |
| C1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)Cl |
| C13H9ClO2 | |
| MFCD00040854 | |
| benzoic acid, 4-chlorophenyl ester, p-chlorophenol benzoate, p-chlorophenyl benzoate, benzoic acid 4-chlorophenyl ester, benzoic acid, p-chlorophenyl ester, 4-chlorophenylbenzoate, acmc-1cina, cbmicro_005244, 4-chlorophenyl benzoate | |
| (4-chlorophenyl) benzoate |
Safety and Handling
TSCA : No