missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenoxyacetic Acid 98.0+%, TCI America™

Catalog No. C025025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C025025G 25 g
C0250500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C025025G Supplier TCI America Supplier No. C025025G

Chemical Identifiers

CAS 122-88-3
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00004305
InChI Key SODPIMGUZLOIPE-UHFFFAOYSA-N
Synonym 4-chlorophenoxyacetic acid, 2-4-chlorophenoxy acetic acid, 4-chlorophenoxy acetic acid, 4-cpa, sure-set, tomatotone, tomato fix, acetic acid, 4-chlorophenoxy, p-chlorophenoxyacetic acid, tomato hold
PubChem CID 26229
ChEBI CHEBI:1808
IUPAC Name 2-(4-chlorophenoxy)acetic acid
SMILES C1=CC(=CC=C1OCC(=O)O)Cl

Specifications

Melting Point 158°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 186.59
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material 4-Chlorophenoxyacetic Acid
EINECSNumber (9)-0423
RTECSNumber AG0175000
TSCA Yes