missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenoxyacetic acid, 98%, Thermo Scientific™

Catalog No. AC166961000
Change view
Click to view available options
100g; Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC166961000 100g; Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC166961000 Supplier Thermo Scientific Chemicals Supplier No. 166961000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 122-88-3
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Molecular Formula C8H7ClO3
Linear Formula ClC6H4OCH2CO2H
Quantity 100g
MDL Number MFCD00004305
Beilstein 06, 187
Synonym 4-chlorophenoxyacetic acid, 2-4-chlorophenoxy acetic acid, 4-chlorophenoxy acetic acid, 4-cpa, sure-set, tomatotone, tomato fix, acetic acid, 4-chlorophenoxy, p-chlorophenoxyacetic acid, tomato hold
Solubility Information Solubility in water: sparingly soluble. Other solubilities: soluble in most common organic solvents
InChI Key SODPIMGUZLOIPE-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1OCC(=O)O)Cl
IUPAC Name 2-(4-chlorophenoxy)acetic acid
Molecular Weight (g/mol) 186.59
PubChem CID 26229
ChEBI CHEBI:1808
Formula Weight 186.59
Percent Purity 98%
Chemical Name or Material 4-Chlorophenoxyacetic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-581-3
RTECSNumber AG0175000
TSCA TSCA

RUO – Research Use Only