missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenacyl Chloride 98.0+%, TCI America™

Catalog No. C13065G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13065G 5 g
C130625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13065G Supplier TCI America Supplier No. C13065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 937-20-2
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.035
MDL Number MFCD00018926
InChI Key FWDFNLVLIXAOMX-UHFFFAOYSA-N
Synonym 2,4'-dichloroacetophenone, 2-chloro-1-4-chlorophenyl ethanone, 4-chlorophenacyl chloride, 2-chloro-1-4-chlorophenyl ethan-1-one, ethanone, 2-chloro-1-4-chlorophenyl, alpha,4-dichloroacetophenone, p-chlorophenacyl chloride, pubchem3372, 2,4-dichloroacetophenone, acmc-209rn9
PubChem CID 70298
IUPAC Name 2-chloro-1-(4-chlorophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CCl)Cl

Specifications

Melting Point 101°C
Color White
Boiling Point 270°C
Quantity 5 g
Formula Weight 189.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorophenacyl Chloride