missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Chloromethyl)thiazole Hydrochloride 98.0+%, TCI America™
$102.52 - $331.81
Chemical Identifiers
| CAS | 7709-58-2 |
|---|---|
| Molecular Formula | C4H5Cl2NS |
| Molecular Weight (g/mol) | 170.05 |
| MDL Number | MFCD02094738 |
| InChI Key | NVTBASMQHFMANH-UHFFFAOYSA-N |
| PubChem CID | 2763289 |
| IUPAC Name | hydrogen 4-(chloromethyl)-1,3-thiazole chloride |
| SMILES | [H+].[Cl-].ClCC1=CSC=N1 |
Chemical Identifiers
| 7709-58-2 | |
| 170.05 | |
| NVTBASMQHFMANH-UHFFFAOYSA-N | |
| hydrogen 4-(chloromethyl)-1,3-thiazole chloride |
| C4H5Cl2NS | |
| MFCD02094738 | |
| 2763289 | |
| [H+].[Cl-].ClCC1=CSC=N1 |
Specifications
| 7709-58-2 | |
| C4H5Cl2NS | |
| 5 g | |
| [H+].[Cl-].ClCC1=CSC=N1 | |
| 170.05 | |
| 170.05 | |
| Crystalline Powder |
| Yellow | |
| MFCD02094738 | |
| NVTBASMQHFMANH-UHFFFAOYSA-N | |
| hydrogen 4-(chloromethyl)-1,3-thiazole chloride | |
| 2763289 | |
| ≥98.0% (HPLC,T) | |
| 4-(Chloromethyl)thiazole Hydrochloride |
Safety and Handling
TSCA : No