missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)thiazole Hydrochloride 98.0+%, TCI America™

Catalog No. C16235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16235G 5 g
C162325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16235G Supplier TCI America Supplier No. C16235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7709-58-2
Molecular Formula C4H5Cl2NS
Molecular Weight (g/mol) 170.05
MDL Number MFCD02094738
InChI Key NVTBASMQHFMANH-UHFFFAOYSA-N
PubChem CID 2763289
IUPAC Name hydrogen 4-(chloromethyl)-1,3-thiazole chloride
SMILES [H+].[Cl-].ClCC1=CSC=N1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 170.05
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Chloromethyl)thiazole Hydrochloride