missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Chloromethyl)-4'-methyl-2,2'-bipyridyl 97.0+%, TCI America™
Supplier: TCI America C33291G
Specifications
| 4-(Chloromethyl)-4′-methyl-2,2′-bipyridyl | |
| 57°C | |
| MFCD24464190 | |
| 3261 | |
| UHTKTJKJDJOOKE-UHFFFAOYSA-N | |
| 2-[4-(chloromethyl)pyridin-2-yl]-4-methylpyridine | |
| 57285230 | |
| ≥97.0% (GC) |
| 83799-54-6 | |
| C12H11ClN2 | |
| 1 g | |
| 4-(Chloromethyl)-4′C-methyl-2,2′C-bipyridine | |
| CC1=CC(=NC=C1)C2=NC=CC(=C2)CCl | |
| 218.684 | |
| 218.68 | |
| Crystalline Powder |
Chemical Identifiers
| 83799-54-6 | |
| 218.684 | |
| UHTKTJKJDJOOKE-UHFFFAOYSA-N | |
| 57285230 | |
| CC1=CC(=NC=C1)C2=NC=CC(=C2)CCl |
| C12H11ClN2 | |
| MFCD24464190 | |
| 4-(Chloromethyl)-4′C-methyl-2,2′C-bipyridine | |
| 2-[4-(chloromethyl)pyridin-2-yl]-4-methylpyridine |
Safety and Handling
TSCA : No