missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)-4'-methyl-2,2'-bipyridyl 97.0+%, TCI America™

Catalog No. C3329200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3329200MG 200 mg
C33291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3329200MG Supplier TCI America Supplier No. C3329200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 83799-54-6
Molecular Formula C12H11ClN2
Molecular Weight (g/mol) 218.684
MDL Number MFCD24464190
InChI Key UHTKTJKJDJOOKE-UHFFFAOYSA-N
Synonym 4-(Chloromethyl)-4′C-methyl-2,2′C-bipyridine
PubChem CID 57285230
IUPAC Name 2-[4-(chloromethyl)pyridin-2-yl]-4-methylpyridine
SMILES CC1=CC(=NC=C1)C2=NC=CC(=C2)CCl

Specifications

Melting Point 57°C
Quantity 200 mg
UN Number 3261
Formula Weight 218.68
Percent Purity ≥97.0% (GC)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 4-(Chloromethyl)-4′-methyl-2,2′-bipyridyl